Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271342.1 | NP_001258271.1 | 5 - 6 | 2705 | 59 | |
XM_006254228.2 | XP_006254290.1 | 5 - 6 | 2705 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |