Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017443.1 | NP_001017443.1 | 5 - 6 | 660 | 70 | |
XM_006253141.2 | XP_006253203.1 | 5 - 6 | 576 | 70 | ||
XM_006253143.2 | XP_006253205.1 | 6 - 7 | 1657 | 70 | ||
XM_008771257.1 | XP_008769479.1 | 4 - 5 | 462 | 70 | ||
GenBank mRNA | BC092575.1 | - | 5 - 6 | 660 | 7114719 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |