Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcbtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108380.1 | NP_001101850.1 | 8 - 9 | 1414 | 107 | |
XM_008770759.1 | XP_008768981.1 | 9 - 10 | 1240 | 107 | ||
GenBank mRNA | AY724518.1 | - | 7 - 8 | 1060 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |