Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012041.1 | NP_001012041.1 | 6 - 7 | 643 | 68 | |
XM_006251974.2 | XP_006252036.1 | 6 - 7 | 771 | 68 | ||
XM_006251975.1 | XP_006252037.1 | 6 - 7 | 715 | 68 | ||
XM_008770679.1 | XP_008768901.1 | 6 - 7 | 756 | 68 | ||
GenBank mRNA | BC079454.1 | - | 6 - 7 | 643 | 7121327 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |