Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acmsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134372.1 | NP_599199.1 | 101 | 94 | ||
GenBank mRNA | AB069781.1 | - | 101 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |