Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012026.1 | NP_001012026.1 | 5 - 6 | 463 | 101 | |
XM_006249777.2 | XP_006249839.1 | 5 - 6 | 524 | 101 | ||
XM_006249778.2 | XP_006249840.1 | 5 - 6 | 528 | 101 | ||
XM_006249779.2 | XP_006249841.1 | 5 - 6 | 525 | 101 | ||
GenBank mRNA | BC083647.1 | - | 5 - 6 | 463 | 7135053 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |