Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250187.2 | XP_006250249.1 | 2 - 3 | 176 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |