Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057127.1 | NP_476468.1 | 2 - 3 | 347 | 111 | |
XM_006254786.2 | XP_006254848.1 | 2 - 3 | 471 | 111 | ||
GenBank mRNA | D82883.1 | - | 2 - 3 | 347 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |