Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106145.1 | NP_001099615.1 | 3 - 4 | 1068 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Rattus norvegicus Probe spans exons |