Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100514.1 | NP_001093984.1 | 9 - 10 | 1450 | 67 | |
XM_006254858.2 | XP_006254920.1 | 9 - 10 | 1741 | 67 | ||
XM_008772118.1 | XP_008770340.1 | 8 - 9 | 1144 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |