Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109105.1 | NP_001102575.1 | 3 - 4 | 428 | 96 | |
XM_008770891.1 | XP_008769113.1 | 4 - 5 | 342 | 96 | ||
XM_008770892.1 | XP_008769114.1 | 2 - 3 | 335 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |