Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022289.3 | NP_071625.2 | 3 - 4 | 753 | 149 | |
XM_006232135.2 | XP_006232197.1 | 3 - 4 | 766 | 149 | ||
GenBank mRNA | AF068746.2 | - | 2 - 3 | 465 | 149 | |
AF305780.1 | - | 3 - 4 | 739 | 149 | ||
BC061554.1 | - | 3 - 4 | 757 | 6890629 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |