Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl22l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108548.1 | NP_001102018.1 | 3 - 4 | 320 | 125 | |
GenBank mRNA | BC167114.1 | - | 3 - 4 | 237 | 6919691 | 125 |
FQ221224.1 | - | 3 - 4 | 236 | 125 | ||
FQ221258.1 | - | 3 - 4 | 234 | 125 | ||
FQ221490.1 | - | 3 - 4 | 236 | 125 | ||
FQ221726.1 | - | 3 - 4 | 236 | 125 | ||
FQ221812.1 | - | 3 - 4 | 236 | 125 | ||
FQ221852.1 | - | 3 - 4 | 299 | 125 | ||
FQ221904.1 | - | 3 - 4 | 237 | 125 | ||
FQ223463.1 | - | 3 - 4 | 236 | 125 | ||
FQ228314.1 | - | 3 - 4 | 236 | 125 | ||
FQ228345.1 | - | 3 - 4 | 236 | 125 | ||
FQ229046.1 | - | 3 - 4 | 236 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |