Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231949.2 | XP_006232011.1 | 2 - 3 | 674 | 148 | |
GenBank mRNA | AF503609.1 | - | 2 - 3 | 212 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |