Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031724.1 | NP_113912.1 | 5 - 6 | 637 | 84 | |
XM_006232753.2 | XP_006232815.1 | 5 - 6 | 1092 | 84 | ||
XM_006232754.2 | XP_006232816.1 | 5 - 6 | 1142 | 84 | ||
GenBank mRNA | AF295404.1 | - | 2 - 3 | 156 | 84 | |
DV726194.1 | - | 2 - 3 | 178 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |