Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232551.2 | XP_006232613.1 | 5 - 6 | 529 | 75 | |
XM_008761117.1 | XP_008759339.1 | 5 - 6 | 531 | 75 | ||
XM_215595.7 | XP_215595.5 | 4 - 5 | 609 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |