Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017020.3 | NP_058716.2 | 5 - 6 | 813 | 81 | |
XM_006232587.2 | XP_006232649.1 | 5 - 6 | 2247 | 81 | ||
XM_006232588.2 | XP_006232650.1 | 5 - 6 | 2246 | 81 | ||
XM_008761127.1 | XP_008759349.1 | 5 - 6 | 2242 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |