Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107076.1 | NP_001100546.1 | 1 - 2 | 255 | 96 | |
GenBank mRNA | BC168956.1 | - | 1 - 2 | 170 | 7123800 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |