Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109051.1 | NP_001102521.1 | 3 - 4 | 304 | 80 | |
XM_006247907.2 | XP_006247969.1 | 5 - 6 | 1200 | 80 | ||
GenBank mRNA | FQ212712.1 | - | 7 - 8 | 765 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |