Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyzl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135833.1 | NP_001129305.2 | 2 - 3 | 220 | 138 | |
XM_006247503.2 | XP_006247565.1 | 2 - 3 | 1733 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |