Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042561.1 | NP_001036026.1 | 3 - 4 | 576 | 124 | |
XM_008768016.1 | XP_008766238.1 | 2 - 3 | 386 | 124 | ||
GenBank mRNA | BC167746.1 | - | 3 - 4 | 579 | 8364583 | 124 |
CK473503.1 | - | 1 - 2 | 59 | 124 | ||
DY312839.1 | - | 3 - 4 | 578 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |