Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024991.1 | NP_001020162.1 | 1 | 1332 | 127 | |
GenBank mRNA | BC078950.1 | - | 1 | 1332 | 7110058 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |