Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047877.1 | NP_001041342.1 | 2 - 3 | 410 | 124 | |
XM_006247599.2 | XP_006247661.1 | 4 - 5 | 2610 | 124 | ||
XM_006247600.2 | XP_006247662.1 | 3 - 4 | 2467 | 124 | ||
XM_006247601.2 | XP_006247663.1 | 5 - 6 | 2836 | 124 | ||
XM_006247602.2 | XP_006247664.1 | 4 - 5 | 1398 | 124 | ||
XM_006247603.2 | XP_006247665.1 | 4 - 5 | 1252 | 124 | ||
XM_006247604.2 | XP_006247666.1 | 4 - 5 | 2668 | 124 | ||
XM_006247606.2 | XP_006247668.1 | 3 - 4 | 2525 | 124 | ||
XM_006247607.2 | XP_006247669.1 | 3 - 4 | 1437 | 124 | ||
XM_006247608.2 | XP_006247670.1 | 2 - 3 | 1087 | 124 | ||
XM_006247609.2 | XP_006247671.1 | 2 - 3 | 382 | 124 | ||
XM_008768368.1 | XP_008766590.1 | 5 - 6 | 2779 | 124 | ||
XM_008768369.1 | XP_008766591.1 | 6 - 7 | 2243 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |