Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134737.1 | NP_001128209.1 | 2 - 3 | 1615 | 72 | |
GenBank mRNA | BC168206.1 | - | 2 - 3 | 660 | 7130974 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |