Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013118.1 | NP_001013136.1 | 4 - 5 | 841 | 103 | |
XM_008768045.1 | XP_008766267.1 | 4 - 5 | 583 | 103 | ||
XM_008768046.1 | XP_008766268.1 | 4 - 5 | 583 | 103 | ||
GenBank mRNA | BC079102.1 | - | 4 - 5 | 841 | 7124908 | 103 |
FQ225352.1 | - | 4 - 5 | 571 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |