Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012009.1 | NP_001012009.1 | 1 - 2 | 117 | 78 | |
XM_006246958.1 | XP_006247020.1 | 2 - 3 | 253 | 78 | ||
XM_006246959.1 | XP_006247021.1 | 1 - 2 | 441 | 78 | ||
GenBank mRNA | BC088249.1 | - | 1 - 2 | 117 | 7370364 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |