Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pipox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012009.1 | NP_001012009.1 | 6 - 7 | 973 | 70 | |
XM_006246958.1 | XP_006247020.1 | 7 - 8 | 1109 | 70 | ||
GenBank mRNA | BC088249.1 | - | 6 - 7 | 973 | 7370364 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |