Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768311.1 | XP_008766533.1 | 1 - 2 | 144 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |