Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100636.1 | NP_001094106.1 | 5 - 6 | 581 | 89 | |
GenBank mRNA | BC090322.1 | - | 2 - 3 | 248 | 89 | |
L22079.1 | - | 5 - 6 | 501 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |