Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024767.1 | NP_001019938.1 | 2 - 3 | 299 | 109 | |
XM_008769464.1 | XP_008767686.1 | 2 - 3 | 511 | 109 | ||
GenBank mRNA | BC097474.1 | - | 2 - 3 | 299 | 7457808 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |