Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749030.3 | XP_003749078.1 | 3 - 4 | 2887 | 149 | |
XM_006230921.2 | XP_006230983.1 | 4 - 5 | 2396 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |