Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760254.1 | XP_008758476.1 | 1 - 2 | 269 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |