Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifnar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105893.1 | NP_001099363.2 | 9 - 10 | 1555 | 108 | |
NM_001271316.1 | NP_001258245.1 | 10 - 11 | 1745 | 108 | ||
GenBank mRNA | FQ210936.1 | - | 10 - 11 | 1545 | 108 | |
FQ218374.1 | - | 1 - 2 | 176 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons |