Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109023.1 | NP_001102493.1 | 1 - 2 | 135 | 150 | |
GenBank mRNA | FQ234501.1 | - | 1 - 2 | 144 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |