Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008520.1 | NP_001008520.1 | 7 - 8 | 909 | 106 | |
GenBank mRNA | BC086351.1 | - | 7 - 8 | 909 | 7302778 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |