Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105922.1 | NP_001099392.1 | 5 - 6 | 619 | 105 | |
XM_006249131.2 | XP_006249193.1 | 5 - 6 | 638 | 105 | ||
XM_008769130.1 | XP_008767352.1 | 5 - 6 | 638 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |