Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nobox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192013.1 | NP_001178942.1 | 7 - 8 | 970 | 94 | |
XM_008762912.1 | XP_008761134.1 | 7 - 8 | 1165 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |