Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053967.2 | NP_446419.1 | 1 - 2 | 134 | 148 | |
GenBank mRNA | BC081748.1 | - | 1 - 2 | 75 | 7134081 | 148 |
X89999.1 | - | 1 - 2 | 138 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |