Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107851.1 | NP_001101321.1 | 5 - 6 | 587 | 142 | |
XM_006236230.2 | XP_006236292.1 | 3 - 4 | 321 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |