Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capza2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009180.2 | NP_001009180.1 | 5 - 6 | 479 | 167 | |
GenBank mRNA | BC101867.1 | - | 5 - 6 | 479 | 7938377 | 167 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |