Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110365.1 | NP_001103835.1 | 3 - 4 | 692 | 162 | |
GenBank mRNA | D88461.1 | - | 3 - 4 | 398 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |