Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC099166.1 | - | 591 | 7382105 | 67 | |
X14319.1 | - | 285 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |