Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002808.2 | NP_001002808.2 | 9 - 10 | 1457 | 61 | |
XM_006236215.2 | XP_006236277.1 | 10 - 11 | 1996 | 61 | ||
XM_006236216.2 | XP_006236278.1 | 10 - 11 | 1720 | 61 | ||
XM_006236217.2 | XP_006236279.1 | 10 - 11 | 1680 | 61 | ||
XM_006236218.2 | XP_006236280.1 | 10 - 11 | 1251 | 61 | ||
XM_006236219.2 | XP_006236281.1 | 9 - 10 | 2145 | 61 | ||
XM_008762801.1 | XP_008761023.1 | 10 - 11 | 2036 | 61 | ||
XM_008762802.1 | XP_008761024.1 | 10 - 11 | 2012 | 61 | ||
GenBank mRNA | BC085840.1 | - | 9 - 10 | 1457 | 7135802 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |