Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760101.1 | XP_008758323.1 | 1 - 2 | 1011 | 78 | |
XM_008760102.1 | XP_008758324.1 | 1 - 2 | 991 | 78 | ||
XM_008760104.1 | XP_008758326.1 | 1 - 2 | 990 | 78 | ||
GenBank mRNA | FQ212306.1 | - | 1 - 2 | 193 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus 1700 Probe spans exons |