Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106316.1 | NP_001099786.1 | 10 - 11 | 1162 | 123 | |
XM_006230532.2 | XP_006230594.1 | 10 - 11 | 1978 | 123 | ||
GenBank mRNA | BC166496.1 | - | 1107 | 7111527 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |