Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115042.2 | NP_001108514.2 | 7 - 8 | 971 | 65 | |
XM_008760177.1 | XP_008758399.1 | 3 - 4 | 631 | 65 | ||
GenBank mRNA | BC097411.1 | - | 7 - 8 | 901 | 65 | |
BC158663.1 | - | 7 - 8 | 971 | 8369770 | 65 | |
BC158764.1 | - | 7 - 8 | 910 | 7111047 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |