Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsga10ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004278.3 | NP_001004278.2 | 6 - 7 | 1552 | 64 | |
NM_001270737.1 | NP_001257666.1 | 6 - 7 | 1426 | 64 | ||
XM_006230847.2 | XP_006230909.1 | 7 - 8 | 1752 | 64 | ||
XM_008760155.1 | XP_008758377.1 | 7 - 8 | 1762 | 64 | ||
XM_008760156.1 | XP_008758378.1 | 6 - 7 | 1710 | 64 | ||
XM_008760157.1 | XP_008758379.1 | 5 - 6 | 1541 | 64 | ||
GenBank mRNA | BC097254.1 | - | 6 - 7 | 1407 | 7453704 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |