Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sharpin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031153.2 | NP_112415.1 | 6 - 7 | 1258 | 108 | |
GenBank mRNA | AF203906.1 | - | 6 - 7 | 932 | 108 | |
BC083553.1 | - | 6 - 7 | 1258 | 7189111 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |