Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130578.1 | NP_001124050.1 | 1 - 2 | 73 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |