Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053311.1 | NP_445763.1 | 4 - 5 | 929 | 96 | |
XM_006241294.2 | XP_006241356.1 | 4 - 5 | 1021 | 96 | ||
XM_006241295.2 | XP_006241357.1 | 3 - 4 | 898 | 96 | ||
XM_006241296.2 | XP_006241358.1 | 4 - 5 | 1021 | 96 | ||
GenBank mRNA | FQ212562.1 | - | 4 - 5 | 1174 | 96 | |
FQ227803.1 | - | 3 - 4 | 738 | 96 | ||
J03753.1 | - | 4 - 5 | 929 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |